Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024181.2 | NP_077143.2 | 15 | 1978 | ||
GenBank mRNA | AF255459.1 | - | 15 | 1655 | ||
AF314002.1 | - | 14 | 1491 | |||
AK004617.1 | - | 15 | 1733 | |||
AK028184.1 | - | 15 | 1719 | |||
AK128945.1 | - | 15 | 1732 | |||
AK165652.1 | - | 15 | 1735 | |||
AK166502.1 | - | 15 | 1689 | |||
AK169020.1 | - | 15 | 1745 | |||
BC002207.1 | - | 6 | 540 | |||
BC033461.1 | - | 15 | 1717 | 4010341 |