Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039965.1 | NP_001035054.1 | 5 | 1394 | ||
NM_001039967.1 | NP_001035056.1 | 6 | 1502 | |||
NM_181274.3 | NP_851791.1 | 5 | 1462 | |||
GenBank mRNA | AK004570.1 | - | 5 | 1405 | ||
AK030679.1 | - | 6 | 1500 | |||
AK054059.1 | - | 1 | 747 | |||
AK083392.1 | - | 6 | 1417 | |||
AK088889.1 | - | 4 | 1305 | |||
AK136878.1 | - | 5 | 1398 | |||
AK213224.1 | - | 1 | 139 | |||
BC063330.1 | - | 1459 | ||||
BC079868.1 | - | 7 | 1997 |