Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177550.1 | NP_001171021.1 | 4 | 1092 | ||
GenBank mRNA | AB470967.1 | - | 4 | 1099 | ||
BC023805.1 | - | 4 | 1091 | 5352880 |
Gene Symbol: 3300002I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB470968.1 | - | 4 | 645 |