Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025838.2 | NP_080114.1 | 1, 2 | 214 | ||
NM_028336.3 | NP_082612.2 | 1, 2 | 214 | |||
GenBank mRNA | AA217716.1 | - | 1, 2 | 217 | ||
AK002804.1 | - | 1, 2 | 184 | |||
AK003403.1 | - | 1, 2 | 210 | |||
AK012938.1 | - | 1, 2 | 213 | |||
AK154279.1 | - | 1, 2 | 212 | |||
BC096686.1 | - | 1, 2 | 192 | 1446796 | ||
BC120499.1 | - | 1, 2 | 136 | 40129422 | ||
BC137584.1 | - | 1, 2 | 137 | 8860603 |