Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025848.3 | NP_080124.1 | 1 | 75 | ||
GenBank mRNA | AK013503.1 | - | 1 | 40 | ||
AK013962.2 | - | 1 | 43 | |||
AK135970.1 | - | 1 | 35 | |||
BC145731.1 | - | 1 | 35 | 40131034 | ||
BC145733.1 | - | 1 | 35 | 40131036 |