Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025869.3 | NP_080145.1 | 4 | 728 | ||
GenBank mRNA | AB104416.1 | - | 2 | 518 | ||
AK007513.1 | - | 2 | 98 | |||
AK009781.1 | - | 4 | 729 | |||
BC018204.1 | - | 2 | 518 | 4038120 |