Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025870.1 | NP_080146.1 | 2 | 730 | ||
GenBank mRNA | AK009864.1 | - | 2 | 722 | ||
AK043948.1 | - | 2 | 699 | |||
BC019749.1 | - | 2 | 713 | |||
BC150493.1 | - | 2 | 732 | 40142491 |