Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153775.2 | NP_722470.2 | 6 | 521 | ||
GenBank mRNA | AK012440.1 | - | 6 | 486 | ||
AK090349.1 | - | 2 | 116 | |||
AK090391.1 | - | 456 | ||||
AK154995.1 | - | 6 | 456 | |||
AK172353.1 | - | 4 | 964 | |||
BC036332.1 | - | 6 | 474 | 3156706 |