Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025876.2 | NP_080152.1 | 6 | 729 | ||
GenBank mRNA | AK010081.1 | - | 6 | 729 | ||
AK010097.1 | - | 6 | 729 | |||
AK088517.1 | - | 6 | 738 | |||
BC025132.1 | - | 5 | 694 | 3666341 |