Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025886.3 | NP_080162.3 | 3 | 471 | ||
GenBank mRNA | AK002243.1 | - | 3 | 411 | ||
AK010305.1 | - | 3 | 497 | |||
BC011131.1 | - | 2 | 398 | 4206694 | ||
CK617449.1 | - | 3 | 471 |