Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025886.3 | NP_080162.3 | 4 | 973 | ||
GenBank mRNA | AK002243.1 | - | 4 | 913 | ||
AK010305.1 | - | 4 | 999 | |||
AK208797.1 | - | 2 | 123 | |||
AW990538.1 | - | 3 | 554 | |||
BC011131.1 | - | 3 | 900 | 4206694 |