Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025888.5 | NP_080164.2 | 4 | 599 | ||
GenBank mRNA | AK010404.1 | - | 3 | 362 | ||
AK030202.1 | - | 4 | 601 | |||
AK088807.1 | - | 3 | 367 | |||
AK168440.1 | - | 4 | 590 | |||
AK171742.1 | - | 3 | 398 | |||
BC036318.1 | - | 2 | 244 | |||
BC048161.1 | - | 3 | 383 | |||
BC049250.1 | - | 3 | 476 | 5042309 | ||
BC052712.1 | - | 3 | 385 |