Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025891.3 | NP_080167.3 | 12 | 1520 | ||
GenBank mRNA | AK019374.1 | - | 5 | 601 | ||
AK158019.1 | - | 1549 | ||||
BC013122.1 | - | 12 | 1557 | 3586276 | ||
BC060525.1 | - | 12 | 1419 | 6817219 |
Gene Symbol: Chpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138814.1 | - | 4272 |