Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam210b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025912.4 | NP_080188.3 | 2 | 341 | ||
GenBank mRNA | AK008190.1 | - | 2 | 282 | ||
AK076087.1 | - | 2 | 284 | |||
AK084415.1 | - | 2 | 294 | |||
AK147935.1 | - | 2 | 299 | |||
AK148213.1 | - | 2 | 292 | |||
AK148334.1 | - | 2 | 282 | |||
AK148435.1 | - | 2 | 282 | |||
AK149497.1 | - | 2 | 279 | |||
AK160529.1 | - | 2 | 342 | |||
BC016210.1 | - | 2 | 314 | 4924329 | ||
BC057642.1 | - | 2 | 301 | 2655111 |