Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025915.4 | NP_080191.3 | 2 | 291 | ||
GenBank mRNA | AK010789.1 | - | 2 | 298 | ||
AK011240.1 | - | 2 | 292 | |||
AK045149.1 | - | 2 | 292 | |||
AK161789.1 | - | 2 | 292 | |||
BC100316.1 | - | 2 | 280 | |||
BC120912.2 | - | 2 | 292 | 40091214 |