Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025924.2 | NP_080200.1 | 1 | 132 | ||
GenBank mRNA | AK011127.1 | - | 1 | 109 | ||
AK016053.1 | - | 1 | 95 | |||
AK020823.1 | - | 1 | 136 | |||
AK033239.1 | - | 1 | 113 | |||
AK145471.1 | - | 89 | ||||
AK166277.1 | - | 1 | 103 | |||
BC116988.1 | - | 1 | 132 | 40126307 | ||
BC119035.1 | - | 1 | 132 | 8733784 |