Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024188.6 | NP_077150.1 | 7 | 1034 | ||
GenBank mRNA | AB085609.1 | - | 7 | 776 | ||
AK009027.1 | - | 7 | 814 | |||
AK010790.1 | - | 7 | 777 | |||
AK011354.1 | - | 7 | 714 | |||
AK078086.1 | - | 7 | 1034 | |||
AK146158.1 | - | 7 | 792 | |||
AK146342.1 | - | 7 | 779 | |||
AK151662.1 | - | 7 | 773 | |||
AK151695.1 | - | 7 | 764 | |||
AK167681.1 | - | 7 | 864 | |||
BC003422.1 | - | 7 | 738 | 2598824 | ||
EF095212.1 | - | 4 | 427 |