Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025937.4 | NP_080213.3 | 1 | 409 | ||
GenBank mRNA | AK011492.1 | - | 1 | 360 | ||
AK012826.1 | - | 1 | 342 | |||
AK038344.1 | - | 1 | 357 | |||
AK076125.1 | - | 1 | 344 | |||
AK089943.1 | - | 1 | 337 | |||
AY388959.1 | - | 1 | 360 | |||
BC026774.1 | - | 1 | 377 | 4019392 | ||
CJ150721.1 | - | 1 | 410 |