Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291108.1 | NP_001278037.1 | 3 | 495 | ||
NM_025947.3 | NP_080223.2 | 3 | 251 | |||
GenBank mRNA | AK002359.1 | - | 3 | 192 | ||
AK003435.1 | - | 2 | 181 | |||
AK008214.1 | - | 3 | 212 | |||
AK008595.1 | - | 4 | 213 | |||
AK166685.1 | - | 3 | 227 | |||
BC099423.1 | - | 3 | 211 | 30933448 | ||
BC125610.1 | - | 3 | 211 | 40130125 | ||
BC125648.1 | - | 3 | 211 | 40130163 | ||
CJ136086.1 | - | 3 | 496 | |||
CO041603.1 | - | 2 | 448 |