Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 4 | 714 | ||
GenBank mRNA | AB086655.1 | - | 4 | 693 | ||
AF477481.1 | - | 4 | 714 | |||
AK012041.1 | - | 4 | 691 | |||
AK017609.1 | - | 4 | 688 | |||
AK028287.1 | - | 4 | 686 | |||
AK049163.1 | - | 4 | 686 | |||
AK076527.1 | - | 4 | 704 | |||
AK083015.1 | - | 4 | 689 | |||
AK194030.1 | - | 2 | 290 | |||
AK198035.1 | - | 1 | 43 | |||
AK204196.1 | - | 1 | 43 | |||
BC013842.1 | - | 4 | 669 | |||
BC048076.1 | - | 4 | 714 | 6307476 |