Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164557.1 | NP_001158029.1 | 3 | 679 | ||
NM_001164558.1 | NP_001158030.1 | 4 | 376 | |||
NM_026018.3 | NP_080294.1 | 4 | 447 | |||
GenBank mRNA | AB009296.1 | - | 3 | 257 | ||
AK002288.1 | - | 2 | 384 | |||
AK007936.1 | - | 3 | 665 | |||
AK008253.1 | - | 4 | 433 | |||
AK012305.1 | - | 4 | 394 | |||
BC013542.1 | - | 3 | 284 | 4241868 | ||
DV057288.1 | - | 4 | 393 | |||
DV058382.1 | - | 3 | 368 |