Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026036.3 | NP_080312.1 | 1 | 47 | ||
GenBank mRNA | AK012941.1 | - | 1 | 48 | ||
AK030885.1 | - | 1 | 27 | |||
AK128960.1 | - | 1 | 33 | |||
AK145743.1 | - | 1 | 28 | |||
AK150775.1 | - | 1 | 48 | |||
AK151092.1 | - | 1 | 48 | |||
AK151431.1 | - | 1 | 48 | |||
AK152448.1 | - | 1 | 49 | |||
AK152723.1 | - | 1 | 58 | |||
AK152817.1 | - | 1 | 58 | |||
AK153092.1 | - | 1 | 48 | |||
AK154481.1 | - | 1 | 34 | |||
AK159439.1 | - | 1 | 17 | |||
AK159735.1 | - | 1 | 42 | |||
AK165948.1 | - | 1 | 16 | |||
AK167499.1 | - | 1 | 34 | |||
AK167854.1 | - | 1 | 39 | |||
AK170905.1 | - | 1 | 49 | |||
AK171394.1 | - | 1 | 78 | |||
AY241868.1 | - | 1 | 84 | |||
BC027248.1 | - | 1 | 19 | 2811938 |