Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026041.2 | NP_080317.3 | 5 | 791 | ||
GenBank mRNA | AK007616.1 | - | 5 | 745 | ||
AK010658.1 | - | 5 | 759 | |||
AK013217.1 | - | 5 | 759 | |||
AK087997.1 | - | 5 | 761 | |||
AK169088.1 | - | 5 | 792 | |||
BC006787.1 | - | 5 | 757 | 3589453 | ||
CO043575.1 | - | 1 | 434 |