Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026068.2 | NP_080344.2 | 2 | 674 | ||
GenBank mRNA | AK005655.1 | - | 2 | 83 | ||
AK013990.1 | - | 2 | 115 | |||
AK045566.1 | - | 1 | 888 | |||
BC019685.1 | - | 2 | 144 | 3983617 | ||
BC051929.1 | - | 2 | 674 | 5709831 |