Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700037H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026091.2 | NP_080367.1 | 4 | 359 | ||
GenBank mRNA | AK003406.1 | - | 4 | 360 | ||
AK006622.1 | - | 4 | 429 | |||
AK040365.1 | - | 1 | 765 | |||
AK048328.1 | - | 1 | 1812 | |||
AK178717.1 | - | 2 | 107 | |||
BC006041.1 | - | 3 | 271 | 3497476 | ||
BC071212.1 | - | 5 | 475 |