Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026092.3 | NP_080368.1 | 5 | 569 | ||
GenBank mRNA | AK006241.1 | - | 5 | 570 | ||
AK006630.1 | - | 5 | 570 | |||
AY572447.1 | - | 5 | 528 | |||
BC048450.1 | - | 5 | 540 | 6774504 |