Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039951.2 | NP_001035040.1 | 4 | 561 | ||
NM_026112.4 | NP_080388.2 | 4 | 561 | |||
GenBank mRNA | AK010575.1 | - | 4 | 556 | ||
AK021179.1 | - | 4 | 512 | |||
AK032301.1 | - | 4 | 564 | |||
AK039817.1 | - | 4 | 507 | |||
AK044645.1 | - | 4 | 529 | |||
AK139269.1 | - | 4 | 562 | |||
AK143200.1 | - | 4 | 526 | |||
AK165095.1 | - | 4 | 500 | |||
AK173278.1 | - | 4 | 503 | |||
BC052747.1 | - | 4 | 480 | 6837952 | ||
BC110665.1 | - | 3 | 171 |