Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026132.2 | NP_080408.2 | 4 | 328 | ||
GenBank mRNA | AK006683.1 | - | 4 | 308 | ||
AK015240.1 | - | 4 | 329 | |||
AY495589.1 | - | 4 | 233 | |||
BC049564.1 | - | 4 | 299 | |||
BC116763.1 | - | 4 | 329 | 40126082 | ||
BC116765.1 | - | 4 | 329 | 40126084 | ||
BC144839.1 | - | 4 | 393 | |||
BC144840.1 | - | 3 | 269 |