Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026160.4 | NP_080436.1 | 3 | 237 | ||
GenBank mRNA | AF255953.1 | - | 3 | 255 | ||
AK002795.1 | - | 3 | 240 | |||
AK003106.1 | - | 3 | 238 | |||
AK003205.1 | - | 3 | 238 | |||
AK003558.1 | - | 3 | 240 | |||
AK012604.1 | - | 3 | 241 | |||
AK132329.1 | - | 245 | ||||
AK151614.1 | - | 3 | 236 | |||
BC068180.1 | - | 3 | 240 | 30634082 | ||
BC086774.1 | - | 2 | 2664 | |||
BC110308.1 | - | 3 | 228 | 5319360 |