Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253390.1 | NP_001240319.1 | 5, 6 | 646 | ||
NM_001253391.1 | NP_001240320.1 | 5, 6 | 808 | |||
NM_026182.5 | NP_080458.1 | 5, 6 | 656 | |||
GenBank mRNA | AF354708.1 | - | 4, 5 | 607 | ||
AK004846.1 | - | 5, 6 | 634 | |||
AK006952.1 | - | 5, 6 | 808 | |||
AK009195.2 | - | 5, 6 | 622 | |||
AK012823.1 | - | 634 | ||||
AK016316.1 | - | 5, 6 | 796 | |||
AK018048.1 | - | 5, 6 | 638 | |||
AK075822.1 | - | 5, 6 | 632 | |||
AK075992.1 | - | 5, 6 | 656 | |||
AK134822.1 | - | 6, 7 | 650 | |||
AK161251.1 | - | 5, 6 | 629 | |||
AK193362.1 | - | 2, 3 | 298 | |||
BC019971.1 | - | 5, 6 | 604 | 4159093 | ||
BC042471.1 | - | 4, 5 | 580 | 3988195 | ||
DQ369744.1 | - | 5, 6 | 767 |