Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253390.1 | NP_001240319.1 | 6 | 741 | ||
NM_001253391.1 | NP_001240320.1 | 6 | 903 | |||
NM_026182.5 | NP_080458.1 | 6 | 751 | |||
GenBank mRNA | AF354708.1 | - | 5 | 702 | ||
AK004846.1 | - | 6 | 729 | |||
AK006952.1 | - | 6 | 903 | |||
AK009195.2 | - | 6 | 717 | |||
AK012823.1 | - | 729 | ||||
AK016316.1 | - | 6 | 891 | |||
AK075822.1 | - | 6 | 727 | |||
AK075992.1 | - | 6 | 751 | |||
AK134822.1 | - | 7 | 745 | |||
AK161251.1 | - | 6 | 724 | |||
BC019971.1 | - | 6 | 699 | 4159093 | ||
BC042471.1 | - | 5 | 675 | 3988195 | ||
DQ369744.1 | - | 6 | 862 |