Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023348.4 | NP_075837.3 | 5 | 855 | ||
GenBank mRNA | AK005042.1 | - | 5 | 805 | ||
AK007065.1 | - | 5 | 848 | |||
AK039203.1 | - | 5 | 824 | |||
AK147845.1 | - | 5 | 763 | |||
AY008722.1 | - | 5 | 852 | |||
BC030066.1 | - | 5 | 804 | 5118167 |