Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025615.3 | NP_001020786.1 | 4, 5 | 464 | ||
NM_001289436.1 | NP_001276365.1 | 5, 6 | 510 | |||
NM_026202.4 | NP_080478.2 | 4, 5 | 464 | |||
GenBank mRNA | AJ534985.1 | - | 4, 5 | 331 | ||
AK012190.1 | - | 4, 5 | 358 | |||
AK016827.1 | - | 4, 5 | 439 | |||
AK077557.1 | - | 4, 5 | 402 | |||
AK147820.1 | - | 4, 5 | 415 | |||
AK151672.1 | - | 4, 5 | 426 | |||
AK162434.1 | - | 4, 5 | 441 | |||
AK165214.1 | - | 4, 5 | 369 | |||
BC144895.1 | - | 4, 5 | 443 | 9053443 | ||
BC144898.1 | - | 5, 6 | 490 | |||
BC145933.1 | - | 4, 5 | 443 | 40131236 | ||
BC145935.1 | - | 4, 5 | 443 | 40131238 | ||
BY716501.1 | - | 4, 5 | 439 | |||
CJ045424.1 | - | 4, 5 | 468 |