Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026232.3 | NP_080508.1 | 2, 3 | 238 | ||
GenBank mRNA | AK015679.1 | - | 2, 3 | 239 | ||
AK017035.1 | - | 2, 3 | 239 | |||
AK030950.1 | - | 2, 3 | 207 | |||
AK090086.1 | - | 2, 3 | 176 | |||
AK151334.1 | - | 2, 3 | 207 | |||
AK159122.1 | - | 2, 3 | 236 | |||
AK159732.1 | - | 2, 3 | 236 | |||
BC022676.1 | - | 2, 3 | 178 | 4238839 |