Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026261.2 | NP_080537.1 | 1 | 558 | ||
GenBank mRNA | AK006408.1 | - | 1 | 484 | ||
AK015868.1 | - | 1 | 559 | |||
AK016746.1 | - | 1 | 515 | |||
BC048443.1 | - | 1 | 497 | 6775231 | ||
BC061071.1 | - | 1 | 523 | 6774419 |