Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: l7Rn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291286.1 | NP_001278215.1 | 4 | 656 | ||
NM_001291287.1 | NP_001278216.1 | 5 | 780 | |||
NM_001291288.1 | NP_001278217.1 | 5 | 857 | |||
NM_001291289.1 | NP_001278218.1 | 3 | 478 | |||
NM_026304.3 | NP_080580.1 | 4 | 716 | |||
GenBank mRNA | AK002306.1 | - | 4 | 698 | ||
AK003324.1 | - | 3 | 469 | |||
AK008604.1 | - | 3 | 480 | |||
AK012166.1 | - | 4 | 682 | |||
BC003916.1 | - | 4 | 664 | 3485003 | ||
BC083330.1 | - | 5 | 718 | |||
CD776674.1 | - | 2 | 127 | |||
CO038997.1 | - | 2 | 366 | |||
DQ196315.1 | - | 4 | 694 | |||
DQ196316.1 | - | 4 | 694 |