Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 3 | 285 | ||
NM_026948.3 | NP_081224.1 | 2 | 233 | |||
GenBank mRNA | AA197830.1 | - | 2 | 235 | ||
AK002828.1 | - | 3 | 229 | |||
AK003381.1 | - | 2 | 173 | |||
AK003647.1 | - | 3 | 227 | |||
AK004197.1 | - | 3 | 227 | |||
AK007580.1 | - | 2 | 253 | |||
AK007912.1 | - | 2 | 276 | |||
AK012608.1 | - | 2 | 492 | |||
AK145185.1 | - | 278 | ||||
AK159336.1 | - | 3 | 232 | |||
BC021463.1 | - | 1 | 352 | |||
BC024422.1 | - | 3 | 215 | 5005082 | ||
BC028531.1 | - | 3 | 215 | |||
BC037500.1 | - | 1 | 211 | |||
BY033979.1 | - | 2 | 287 | |||
BY057392.1 | - | 2 | 252 | |||
CA323205.1 | - | 2 | 199 | |||
CK624897.1 | - | 2 | 210 |