Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026307.3 | NP_080583.3 | 3, 4 | 356 | ||
NM_026948.3 | NP_081224.1 | 2, 3 | 304 | |||
GenBank mRNA | AA197830.1 | - | 2, 3 | 306 | ||
AK002828.1 | - | 3, 4 | 300 | |||
AK003381.1 | - | 2, 3 | 244 | |||
AK003647.1 | - | 3, 4 | 298 | |||
AK004197.1 | - | 3, 4 | 298 | |||
AK007580.1 | - | 2, 3 | 324 | |||
AK007912.1 | - | 2, 3 | 347 | |||
AK012608.1 | - | 2, 3 | 563 | |||
AK159336.1 | - | 3, 4 | 303 | |||
AK181681.1 | - | 1, 2 | 78 | |||
AK187338.1 | - | 1, 2 | 223 | |||
BC024422.1 | - | 3, 4 | 286 | 5005082 | ||
BC028531.1 | - | 3, 4 | 286 | |||
BC037500.1 | - | 1, 2 | 282 | |||
BY033979.1 | - | 2, 3 | 358 | |||
BY057392.1 | - | 2, 3 | 323 | |||
CA323205.1 | - | 2, 3 | 270 | |||
CK624897.1 | - | 2, 3 | 281 |