Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hypk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026318.3 | NP_080594.2 | 1 | 151 | ||
GenBank mRNA | AK003580.1 | - | 1 | 145 | ||
AK009109.1 | - | 1 | 139 | |||
AK168506.1 | - | 1 | 120 | |||
BC021588.1 | - | 1 | 115 | 5116361 | ||
BC038469.1 | - | 1 | 153 | 5028838 | ||
BC060244.1 | - | 1 | 546 | |||
BY165803.1 | - | 1 | 153 |