Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hypk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026318.3 | NP_080594.2 | 4 | 390 | ||
GenBank mRNA | AI847440.1 | - | 1 | 227 | ||
AK003580.1 | - | 4 | 384 | |||
AK009109.1 | - | 4 | 378 | |||
AK168506.1 | - | 4 | 359 | |||
BC021588.1 | - | 3 | 644 | 5116361 | ||
BC038469.1 | - | 4 | 392 | 5028838 | ||
BC060244.1 | - | 3 | 1139 |