Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ribc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026357.2 | NP_080633.2 | 7 | 1272 | ||
GenBank mRNA | AK006801.1 | - | 8 | 1381 | ||
AK016311.1 | - | 7 | 1273 | |||
AK017028.1 | - | 7 | 1291 | |||
BC112373.1 | - | 7 | 1197 | 40056860 |