Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026360.3 | NP_080636.2 | 2 | 181 | ||
GenBank mRNA | AK010310.1 | - | 2 | 174 | ||
AK076982.1 | - | 2 | 166 | |||
BC060168.1 | - | 2 | 161 | |||
BC095944.1 | - | 2 | 158 | 4935970 | ||
BC132242.1 | - | 2 | 173 | 40130519 | ||
BC132244.1 | - | 2 | 173 | 40130521 | ||
BY123864.1 | - | 2 | 181 |