Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plgrkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026362.2 | NP_080638.1 | 5 | 430 | ||
GenBank mRNA | AK017215.1 | - | 5 | 431 | ||
AK168454.1 | - | 5 | 406 | |||
AK170700.1 | - | 6 | 504 | |||
BC024953.1 | - | 5 | 389 | 4951101 |