Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175121.3 | NP_780330.2 | 10 | 1209 | ||
GenBank mRNA | AK031615.1 | - | 10 | 1240 | ||
AK129342.1 | - | 10 | 1170 | |||
AK164568.1 | - | 4 | 4972 | |||
BC037675.1 | - | 7 | 616 | |||
BC041108.1 | - | 10 | 1079 | 4038546 | ||
BC048178.1 | - | 10 | 1209 | 6825801 |