Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175121.3 | NP_780330.2 | 5, 6 | 760 | ||
GenBank mRNA | AK031615.1 | - | 5, 6 | 791 | ||
AK129342.1 | - | 5, 6 | 721 | |||
BC037675.1 | - | 2, 3 | 167 | |||
BC041108.1 | - | 5, 6 | 630 | 4038546 | ||
BC048178.1 | - | 5, 6 | 760 | 6825801 |