Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026399.2 | NP_080675.2 | 5 | 589 | ||
GenBank mRNA | AK005790.1 | - | 5 | 529 | ||
AY365434.1 | - | 5 | 589 | |||
BC005587.1 | - | 4 | 828 | |||
BC019369.1 | - | 5 | 512 | 4014928 | ||
BC020047.1 | - | 1512 | ||||
BC034842.1 | - | 4 | 757 |