Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026417.4 | NP_080693.2 | 5 | 751 | ||
GenBank mRNA | AK009618.1 | - | 5 | 742 | ||
AK017721.1 | - | 5 | 729 | |||
AK017722.1 | - | 5 | 729 | |||
AK083301.1 | - | 5 | 731 | |||
AK159227.1 | - | 5 | 712 | |||
AK166347.1 | - | 5 | 739 | |||
BC003936.1 | - | 6 | 926 | 3486503 |