Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023395.2 | NP_075884.1 | 3 | 518 | ||
GenBank mRNA | AK009971.1 | - | 3 | 521 | ||
AK018575.1 | - | 3 | 379 | |||
AK132541.1 | - | 3 | 519 | |||
AK142707.1 | - | 3 | 387 | |||
AK161606.1 | - | 3 | 384 | |||
BC025850.1 | - | 3 | 334 | |||
BC038122.1 | - | 3 | 360 | |||
BC132073.1 | - | 3 | 447 | 40130350 | ||
BC132469.1 | - | 3 | 447 | 40130746 | ||
BC145305.1 | - | 3 | 447 | 9053859 |