Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141965.1 | NP_001135437.1 | 4 | 328 | ||
NM_026425.3 | NP_080701.1 | 4 | 358 | |||
GenBank mRNA | AK005143.1 | - | 4 | 331 | ||
AK009229.1 | - | 4 | 330 | |||
AK153145.1 | - | 4 | 340 | |||
BC009157.1 | - | 4 | 295 | 2648626 | ||
BC096451.1 | - | 4 | 282 | 3673241 | ||
BC116374.2 | - | 4 | 287 | 40101000 |