Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285452.1 | NP_001272381.1 | 5 | 835 | ||
NM_028975.4 | NP_083251.2 | 6 | 678 | |||
NM_030108.2 | NP_084384.1 | 6 | 678 | |||
GenBank mRNA | AK005493.1 | - | 6 | 570 | ||
AK007607.1 | - | 6 | 486 | |||
AK007750.1 | - | 6 | 567 | |||
AK017281.1 | - | 6 | 547 | |||
AK019127.1 | - | 5 | 504 | |||
AK032555.1 | - | 6 | 585 | |||
AK153441.1 | - | 6 | 503 | |||
AK165369.1 | - | 6 | 480 | |||
AK206266.1 | - | 4 | 298 | |||
BC005562.1 | - | 6 | 479 | 3965864 | ||
BC016570.1 | - | 5 | 551 | 3583937 | ||
BC023966.1 | - | 5 | 544 | 5342147 |